EMAIL THIS PAGE TO A FRIEND
77613 Sigma-Aldrich

Phenol

for molecular biology, ~90% (T), liquid

Synonym: Hydroxybenzene

  • CAS Number 108-95-2

  • Linear Formula C6H5OH

  • Molecular Weight 94.11

  •  Beilstein Registry Number 969616

  •  MDL number MFCD00002143

  •  PubChem Substance ID 329767810

Purchase

Properties

grade   for molecular biology
vapor density   3.24 (vs air)
vapor pressure   0.09 psi ( 55 °C)
  0.36 mmHg ( 20 °C)
InChI Key   ISWSIDIOOBJBQZ-UHFFFAOYSA-N
assay   ~90% (T)
form   liquid
autoignition temp.   1319 °F
expl. lim.   8.6 %
impurities   DNases, none detected
  RNases, none detected
  phosphatases, none detected
  proteases, none detected
  ~10% water
refractive index   n20/D 1.527
bp   182 °C(lit.)
mp   40-42 °C(lit.)
density   1.071 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤5 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg
suitability   suitable for molecular biology
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN 2821 6.1 / PGII
WGK Germany 
2
Flash Point(F) 
174.2 °F
Flash Point(C) 
79 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?