• USA Home
  • 77965 - Tris(2-ethylhexyl)phosphine oxide

EMAIL THIS PAGE TO A FRIEND
77965 Supelco

Tris(2-ethylhexyl)phosphine oxide

Selectophore, ≥95.0%

Synonym: 3-[Bis(2-ethylhexyl)phosphorylmethyl]heptane

  • CAS Number 2785-32-2

  • Empirical Formula (Hill Notation) C24H51OP

  • Molecular Weight 386.63

  •  Beilstein Registry Number 2332446

  •  MDL number MFCD00368279

  •  PubChem Substance ID 329768055

Purchase

Description

Application

Solvent mediator for serotonin selective electrodes

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Making Headway

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?