• USA Home
  • 78087BC - MVP ICON® Temperature Probe

EMAIL THIS PAGE TO A FRIEND
78087BC Millipore

MVP ICON® Temperature Probe

BioControl, Temperature Monitoring for HACCP Management

Purchase

Properties

packaging   pkg of 1 ea
mfr. no.   BioControl Systems, 78087-00
compatibility   for use with Sigma-Aldrich MVP ICON (78300)
storage temp.   room temp

Description

Components

One MVP ICON Temperature Probe

Testing Method

Temperature

Compatibility

Any other combination might cause loss of performance or irreversible damage to both probe and instrument.

Application

Temperature control can be a critical factor in food safety and temperature must be accurately measured and recorded in many HACCP plans. Temperature is one of the most important factors permitting growth of bacteria and limits can be set for various areas of retail food production.

Features and Benefits

Versatile
The MVP ICON Temperature Probe accurately reads temperature across a wide range of applications.

Durable
The rugged, detachable temperature probe makes in-plant testing easy. The stainless steel probe features a pierced tip, allowing for easy penetration into food samples.

General description

The MVP ICON Temperature Probe is a thermistor housed in a stainless steel probe designed for general purpose temperature measurement of liquids or semi-solids ranging from -55 to 125 °C.

System Requirements

• Windows 7 or Windows 10
• Minimum memory requirement 512 MB; recommend at least 1GB or more of RAM
• At least 16 GB of hard disk space
• WXGA video resolution of 1280 x 800 or better
• Mouse or other pointing device
• One available USB port
• Adobe Acrobat Reader or similar PDF Viewer
• Printer (optional)

Suitability

Food Processing, HACCP Monitoring, Sanitation

Legal Information

MVP ICON is a registered trademark of BioControl Systems, Inc.

Storage and Stability

Clean probe thoroughly with water and a mild detergent solution. Rinse with deionized water. Store probe in original packaging when not in use.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Safe - Solutions for Food

MVP ICON
Protocols & Articles

Articles

Automated HACCP Management

With a new year comes new resolutions. We endeavor to set new goals and envision ensuing possibilities for improving performance throughout the facility. With a new year also comes another Hazard Ana...
Keywords: Food Safety, Indicators, Sample preparations, Titrations

Hygiene Monitoring: Rapid and Traditional Methods

When it comes to hygiene monitoring, we provide a diverse range of rapid test solutions that help you easily verify effectiveness of your sanitation program and reduce the risk of contamination in pr...
Keywords: Allergens, Size-exclusion chromatography

MVP ICON® System for ATP Hygiene Monitoring & Management

The MVP ICON® is an instrument and software platform that combines HACCP and hygiene monitoring with powerful program management capabilities.
Keywords: Indicators

Related Content

위생 모니터링을 위한 MVP ICON® System

MVP ICON®은 HACCP 관리기능을 제공하는 최초의 시스템입니다. 최첨단의 PCS (Photon Counting Sensor) 기술로 MVP ICON®은 ATP, pH, 온도, 전도도 및 농도를 측정할 때 뛰어난 정확도, 감도 및 재현성을 제공합니다. 매끄럽고 가벼운 혁신적인 디자인에 사용하기 쉬운 터치스크린이 특징입니다.

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?