• USA Home
  • 78088BC - MVP ICON® pH/Temperature Probe

EMAIL THIS PAGE TO A FRIEND
78088BC Millipore

MVP ICON® pH/Temperature Probe

BioControl, pH Monitoring for HACCP Management

Purchase

Properties

packaging   pkg of 1 ea
mfr. no.   BioControl Systems, 78088-00
compatibility   for use with Sigma-Aldrich MVP ICON (78300)
storage temp.   room temp

Description

Components

One MVP ICON pH/Temperature Probe

Testing Method

pH, Temperature

Compatibility

Any other combination might cause loss of performance or irreversible damage to both probe and instrument.

Application

The monitoring of pH levels during the process of food production is a vital step in the production of high-quality foods. Proper pH not only affects the aesthetic and quality of the product but maintaining a proper pH level is also critical for food safety. The ability to measure pH and temperature simultaneously allows for increased efficiency in the plant.

Features and Benefits

Probe Technology
The MVP ICON pH / Temperature Probe features an Ion Sensitive Field Effect Transistor (ISFET) solid-state pH sensor – the most powerful pH testing technology available.

Increased Efficiency
The probe measures pH and temperature simultaneously for increased efficiency. Advanced algorithms allow for automatic temperature compensation of pH readings across the entire range.

Durable
The rugged, non-glass design allows the probe to be used throughout the entire plant and in the laboratory.

Principle

The probe features a silver / silver chloride / potassium chloride reference system and a thermistor with automatic temperature compensation. Solid-state componentry built into the MVP ICON instrument controls this sensing element.

General description

The MVP ICON pH / Temperature Probe is a semi-conductor designed for general purpose pH measurement of liquids or semi-solids ranging from 0 – 14 pH. Temperature measurements, ranging from 0 – 80 °C, may be taken
simultaneously.

System Requirements

• Windows 7 or Windows 10
• Minimum memory requirement 512 MB; recommend at least 1GB or more of RAM
• At least 16 GB of hard disk space
• WXGA video resolution of 1280 x 800 or better
• Mouse or other pointing device
• One available USB port
• Adobe Acrobat Reader or similar PDF Viewer
• Printer (optional)

Suitability

Food Processing, HACCP Monitoring, Sanitation

Legal Information

MVP ICON is a registered trademark of BioControl Systems, Inc.

Other Notes

Clean probe thoroughly with water, a soft bristled brush (included), and a mild detergent solution. Rinse with deionized water. Leave 1 - 2 drops of deionized water in the black protective cap or clear protective tube, then cover probe tip to prevent dehydration of the reference electrode and allow for faster start-up times.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis


Safe - Solutions for Food

Rapid protein detection test
Protocols & Articles

Articles

Automated HACCP Management

With a new year comes new resolutions. We endeavor to set new goals and envision ensuing possibilities for improving performance throughout the facility. With a new year also comes another Hazard Ana...
Keywords: Food Safety, Indicators, Sample preparations, Titrations

Hygiene Monitoring: Rapid and Traditional Methods

When it comes to hygiene monitoring, we provide a diverse range of rapid test solutions that help you easily verify effectiveness of your sanitation program and reduce the risk of contamination in pr...
Keywords: Allergens, Size-exclusion chromatography

MVP ICON® System for ATP Hygiene Monitoring & Management

The MVP ICON® is an instrument and software platform that combines HACCP and hygiene monitoring with powerful program management capabilities.
Keywords: Indicators

Related Content

위생 모니터링을 위한 MVP ICON® System

MVP ICON®은 HACCP 관리기능을 제공하는 최초의 시스템입니다. 최첨단의 PCS (Photon Counting Sensor) 기술로 MVP ICON®은 ATP, pH, 온도, 전도도 및 농도를 측정할 때 뛰어난 정확도, 감도 및 재현성을 제공합니다. 매끄럽고 가벼운 혁신적인 디자인에 사용하기 쉬운 터치스크린이 특징입니다.

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?