• USA Home
  • 78089BC - MVP ICON® Conductivity Probe

EMAIL THIS PAGE TO A FRIEND
78089BC Millipore

MVP ICON® Conductivity Probe

BioControl, Sanitizer Concentration and PPM Monitoring for HACCP Management

Purchase

Properties

packaging   pkg of 1 unit
mfr. no.   BioControl Systems, 78089-00
storage condition   room temperature
compatibility   for use with Sigma-Aldrich MVP ICON (78300)

Description

Components

One MVP ICON Conductivity Probe

Testing Method

Conductivity

Compatibility

Any other combination might cause loss of performance or irreversible damage to both probe and instrument. Use of the MVP ICON Dashboard software is required for PPM measurements.

Application

Accurately measuring cleaner and sanitizer concentrations, as parts-per-million (ppm), ensures correct mixing of chemicals prior to application. Excessively high concentrations of these chemicals can result in damaged equipment and thousands of dollars of unnecessary chemical costs. Precise concentration readings ensure chemical solutions are mixed correctly, so they clean and sanitize effectively. Measuring both process controls and CIP rinse water cleanliness provides increased adherence to hygiene and HACCP requirements.

Features and Benefits

Reduce Sanitation Chemical Costs
The MVP ICON Conductivity Probe can accurately measure cleaner and sanitizer concentrations as parts-per-million (ppm), ensuring correct mixing of chemicals prior to application. Excessively high concentrations of these chemicals can result in damaged equipment and thousands of dollars of unnecessary chemical costs. Precise concentration readings ensure chemical solutions are mixed correctly, so they clean and sanitize effectively.

Water Quality Monitoring
The MVP ICON Conductivity Probe measures both process controls and CIP rinse water cleanliness, providing increased adherence to hygiene and HACCP requirements. The probe is safe for food industry use, reusable and requires little maintenance.

Fast and Easy Reporting
The MVP ICON Conductivity Probe measures conductivity, concentration, and temperature simultaneously, saving time and labor. Results are automatically recorded in the MVP ICON database for quick upload to PC, eliminating manual record keeping.

Principle

The Conductivity Probe uses a 4-graphite electrode cell design (K=0.45) with a 1 cm cell length featuring a thermistor to provide temperature readings, ranging from 0 – 80 °C, with automatic temperature compensation. Solid-state componentry built into the MVP ICON instrument controls this sensing element.

General description

The MVP ICON Conductivity Probe is designed for general purpose measurements of Conductivity (μS/cm), Concentration (ppm), and Temperature (°C or °F) in water-based liquids.

System Requirements

• Windows 7 or Windows 10
• Minimum memory requirement 512 MB; recommend at least 1GB or more of RAM
• At least 16 GB of hard disk space
• WXGA video resolution of 1280 x 800 or better
• Mouse or other pointing device
• One available USB port
• Adobe Acrobat Reader or similar PDF Viewer
• Printer (optional)

Suitability

Food Processing, HACCP Monitoring, Sanitation

Legal Information

MVP ICON is a registered trademark of BioControl Systems, Inc.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Safe - Solutions for Food

MVP ICON
Protocols & Articles

Articles

Automated HACCP Management

With a new year comes new resolutions. We endeavor to set new goals and envision ensuing possibilities for improving performance throughout the facility. With a new year also comes another Hazard Ana...
Keywords: Food Safety, Indicators, Sample preparations, Titrations

Hygiene Monitoring: Rapid and Traditional Methods

When it comes to hygiene monitoring, we provide a diverse range of rapid test solutions that help you easily verify effectiveness of your sanitation program and reduce the risk of contamination in pr...
Keywords: Allergens, Size-exclusion chromatography

MVP ICON® System for ATP Hygiene Monitoring & Management

The MVP ICON® is an instrument and software platform that combines HACCP and hygiene monitoring with powerful program management capabilities.
Keywords: Indicators

Related Content

위생 모니터링을 위한 MVP ICON® System

MVP ICON®은 HACCP 관리기능을 제공하는 최초의 시스템입니다. 최첨단의 PCS (Photon Counting Sensor) 기술로 MVP ICON®은 ATP, pH, 온도, 전도도 및 농도를 측정할 때 뛰어난 정확도, 감도 및 재현성을 제공합니다. 매끄럽고 가벼운 혁신적인 디자인에 사용하기 쉬운 터치스크린이 특징입니다.

Related Products

related product

Product #

Description

Add to Cart

78300BC MVP ICON, BioControl, ICON ATP Monitoring for HACCP Management
06999 Conductivity Standard, conductivity 500 μS/cm at 25 °C, ≤2.5% KCl (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?