EMAIL THIS PAGE TO A FRIEND
78104 Millipore

Nutrient Broth No 5

for microbiology

Synonym: Nutrient Broth with 1% Peptone

Purchase

Properties

Related Categories All Dairy Media (alphabetical), All Liquid Media/Broths, Campylobacter, Clinical Media, Dairy Quality Control Media,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   beef extract, 10 g/L
  peptic digest of animal tissue, 10 g/L
  sodium chloride, 5 g/L
Featured Industry   Clinical Testing
Environmental
Food and Beverages
suitability   nonselective for Enterobacter
  nonselective for Escherichia coli and Coliforms
  nonselective for Klebsiella
  nonselective for Salmonella
  selective for Campylobacter

Description

Application

A nutritious medium suitable for the cultivation of fastidious microorganisms.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?