EMAIL THIS PAGE TO A FRIEND
79257 Supelco

Tiliroside

analytical standard

Synonym: Astragalin 6"-trans-p-coumarate, Kaempferol 3-O-β-D-(6″-O-(E)-p-coumaroyl)glucopyranoside, Potengriffioside A

  • CAS Number 20316-62-5

  • Empirical Formula (Hill Notation) C30H26O13

  • Molecular Weight 594.52

  •  MDL number MFCD00017454

  •  PubChem Substance ID 329768345

  •  NACRES NA.24

Purchase

Properties

Related Categories Alphabetical Index of Phytochemical Standards, Analytical Standards, Analytical/Chromatography, Flavonoids, Phytochemical Standards,
Quality Level   100
grade   analytical standard
assay   ≥98% (HPLC)
form   neat
optical activity   [α]/D -65±8°, c = 0.1 in methanol
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Food and Beverages
format   neat
storage temp.   2-8°C
SMILES string   O[C@H]1[C@H](O)[C@@H](COC(=O)\C=C\c2ccc(O)cc2)O[C@@H](OC3=C(Oc4cc(O)cc(O)c4C3=O)c5ccc(O)cc5)[C@@H]1O
InChI   1S/C30H26O13/c31-16-6-1-14(2-7-16)3-10-22(35)40-13-21-24(36)26(38)27(39)30(42-21)43-29-25(37)23-19(34)11-18(33)12-20(23)41-28(29)15-4-8-17(32)9-5-15/h1-12,21,24,26-27,30-34,36,38-39H,13H2/b10-3+/t21-,24-,26+,27-,30+/m1/s1
InChI key   DVGGLGXQSFURLP-VWMSDXGPSA-N

Description

General description

Tiliroside is a glycosidic flavonoid isolated from Helichrysum italicum. It exhibits a variety of pharmacological properties such as anti-inflammatory, antioxidant, anticarcinogenic and hepatoprotective activity.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Tiliroside may be used as a reference standard in the separation and quantification of tiliroside from plant extracts using solid phase extraction (SPE) followed by reversed-phase high-performance liquid chromatography (RP-HPLC).

Packaging

10 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?