EMAIL THIS PAGE TO A FRIEND
80488 Supelco

R-(+)-Methysticin

analytical standard

Synonym: (R)-6-[(E)-2-(1,3-Benzodioxol-5-yl)ethenyl]-5,6-dihydro-4-methoxy-2H-pyran-2-one, d-Methysticin, Kavahin, Kavatin

  • CAS Number 495-85-2

  • Empirical Formula (Hill Notation) C15H14O5

  • Molecular Weight 274.27

  •  Beilstein/REAXYS Number 91701

  •  MDL number MFCD21364778

  •  PubChem Substance ID 329768904

  •  NACRES NA.24

Purchase

Properties

Related Categories Alphabetical Index of Phytochemical Standards, Analytical Standards, Analytical/Chromatography, I - M, Phenylpropanes,
Quality Level   100
grade   analytical standard
assay   ≥98.0% (HPLC)
form   neat
optical activity   [α]/D 118±2°, c = 0.1 in dichloromethane
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤5.0% water
Featured Industry   Food and Beverages
format   neat
storage temp.   2-8°C
SMILES string   COC1=CC(=O)O[C@H](C1)\C=C\c2ccc3OCOc3c2
InChI   1S/C15H14O5/c1-17-12-7-11(20-15(16)8-12)4-2-10-3-5-13-14(6-10)19-9-18-13/h2-6,8,11H,7,9H2,1H3/b4-2+/t11-/m0/s1
InChI key   GTEXBOVBADJOQH-FWEMWIAWSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?