• USA Home
  • 80699 - (−)-Myrtenyl acetate

EMAIL THIS PAGE TO A FRIEND
80699 Supelco

(−)-Myrtenyl acetate

analytical standard

Synonym: (1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-methyl acetate

  • CAS Number 36203-31-3

  • Empirical Formula (Hill Notation) C12H18O2

  • Molecular Weight 194.27

  •  Beilstein Registry Number 4309977

  •  EC Number 252-909-9

  •  MDL number MFCD00126976

  •  PubChem Substance ID 329769038

Purchase

Properties

Related Categories All Alphabetically Sorted, Analytical Standards, Analytical/Chromatography, Flavors and Fragrance Standards, Food & Beverage Standards,
grade   analytical standard
InChI Key   BKATZVAUANSCKN-UHFFFAOYSA-N
assay   ≥95.0% (GC)
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.470-1.474
  n20/D 1.472(lit.)
bp   134-135 °C/38 mmHg(lit.)
density   0.991 g/mL at 25 °C(lit.)
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?