• USA Home
  • 84475 - Sandalwood oil from Santalum album L.

EMAIL THIS PAGE TO A FRIEND
84475 Sigma-Aldrich

Sandalwood oil from Santalum album L.

natural

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, Q-Z More...
quality   natural
refractive index   n20/D 1.50
Organoleptic   woody
storage temp.   2-8°C
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?