EMAIL THIS PAGE TO A FRIEND
86352 Millipore

TBG Broth

for microbiology

Synonym: Tetrathionate Brilliant Green Bile Enrichment Broth

  •  NACRES NA.74

Purchase

Properties

Related Categories All Food Control Media (alphabetical), All Liquid Media/Broths, Clinical Media, Media by Application, Media by Microorganism,
Quality Level   100
grade   for microbiology
form   powder
shelf life   limited shelf life, expiry date on the label
composition   brilliant green, 0.07 g/L
  calcium carbonate, 20 g/L
  meat peptone, 8.6 g/L
  ox-bile (dried), 8 g/L
  potassium tetrathionate, 20 g/L
  sodium chloride, 6.4 g/L
application(s)   microbiological culture: suitable
  pathogen testing: suitable
  sample preparation: suitable
final pH   7.0±0.2 (25 °C)
Featured Industry   Clinical Testing
Environmental
Food and Beverages
suitability   selective for Salmonella spp.

Description

Application

For the selective enrichment of Salmonella species from feces, food and meat samples etc.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Differentiation and Identification Media for Salmonella

Salmonella contamination is the second leading cause of food-borne illness worldwide. Controlling outbreaks of Salmonella is an important task for food regulators, restaurants and the food industry i...
Jvo Siegrist
Microbiology Focus Edition 1.3
Keywords: Biochemistry, Fermentation, Food & Beverage, Indicators, Microbiology

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?