• USA Home
  • 86912 - Tetrabutylammonium iodide

EMAIL THIS PAGE TO A FRIEND
86912 Supelco

Tetrabutylammonium iodide

for electrochemical analysis, ≥99.0%

Synonym: TBAI

  • CAS Number 311-28-4

  • Linear Formula (CH3CH2CH2CH2)4N(I)

  • Molecular Weight 369.37

  •  Beilstein/REAXYS Number 3916152

  •  EC Number 206-220-5

  •  MDL number MFCD00011636

  •  PubChem Substance ID 329769351

Purchase

Properties

Related Categories Ammonium Salts, Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Electrochemistry,
grade   for electrochemical analysis
InChI Key   DPKBAXPHAYBPRL-UHFFFAOYSA-M
assay   ≥99.0% (AT)
  ≥99.0%
mp   141-143 °C (lit.)
  144-146 °C
solubility   acetonitrile: 0.1 g/mL, clear, colorless

Description

General description

Tetrabutylammonium iodide (TBAI) is a quaternary ammonium salt, mainly used as electrolyte.

Visit our Sensor Applications portal to learn more.

Application

TBAI has been used as an electrolyte to study its ion-solvent interaction in acetonitrile solution using ATR-FTIR spectroscopy.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BS5450000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?