• USA Home
  • 87729 - Tetramethylammonium hydroxide solution

EMAIL THIS PAGE TO A FRIEND
87729 Sigma-Aldrich

Tetramethylammonium hydroxide solution

~25% in H2O

  • CAS Number 75-59-2

  • Linear Formula (CH3)4N(OH)

  • Molecular Weight 91.15

  •  Beilstein Registry Number 3558708

  •  MDL number MFCD00008280

  •  PubChem Substance ID 329769603

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Organic Bases, Synthetic Reagents More...
InChI Key   WGTYBPLFGIVFAS-UHFFFAOYSA-M
concentration   ~25% in H2O
impurities   ≤0.1% halides (as chloride)
refractive index   n20/D 1.382
anion traces   sulfate (SO42-): ≤1000 mg/kg

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1835 8 / PGII
WGK Germany 
1
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?