• USA Home
  • 88104 - Tetrapropylammonium bromide

EMAIL THIS PAGE TO A FRIEND
88104 Supelco

Tetrapropylammonium bromide

for electrochemical analysis, ≥99.0%

  • CAS Number 1941-30-6

  • Linear Formula (CH3CH2CH2)4N(Br)

  • Molecular Weight 266.26

  •  Beilstein/REAXYS Number 3567846

  •  EC Number 217-727-6

  •  MDL number MFCD00011840

  •  PubChem Substance ID 329769632

  •  NACRES NB.61

Purchase

Properties

Related Categories Ammonium Salts, Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Electrochemistry,
Quality Level   100
grade   for electrochemical analysis
assay   ≥99.0% (AT)
  ≥99.0%
solubility   acetonitrile: 0.1 g/mL, clear, colorless
SMILES string   [Br-].CCC[N+](CCC)(CCC)CCC
InChI   1S/C12H28N.BrH/c1-5-9-13(10-6-2,11-7-3)12-8-4;/h5-12H2,1-4H3;1H/q+1;/p-1
InChI key   BGQMOFGZRJUORO-UHFFFAOYSA-M

Description

General description

Visit our Sensor Applications portal to learn more.

Application

Supporting electrolyte

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?