EMAIL THIS PAGE TO A FRIEND
88860 Sigma-Aldrich

p-Thiocresol

purum, ≥98.0% (RT)

Synonym: 4-Mercaptotoluene, 4-Methylthiophenol, p-Toluenethiol, p-Tolyl mercaptan

  • CAS Number 106-45-6

  • Linear Formula CH3C6H4SH

  • Molecular Weight 124.20

  •  Beilstein Registry Number 605761

  •  EC Number 203-399-1

  •  MDL number MFCD00004851

Purchase

Properties

vapor density   6.2 (vs air)
vapor pressure   10 mmHg ( 71 °C)
InChI Key   WLHCBQAPPJAULW-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (RT)
bp   195 °C(lit.)
mp   41-43 °C (lit.)
  41-43 °C

Description

Other Notes

This product has been replaced by T28525-ALDRICH | 4-Methylbenzenethiol 98%

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3335
WGK Germany 
3
RTECS 
XT8925000
Flash Point(F) 
154.4 °F
Flash Point(C) 
68 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

T28525 4-Methylbenzenethiol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?