• USA Home
  • 90006 - Cadmium Standard for ICP

EMAIL THIS PAGE TO A FRIEND
90006 Supelco

Cadmium Standard for ICP

TraceCERT®, 10,000 mg/L Cd in nitric acid

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs),
Quality Level   300
grade   certified reference material
  TraceCERT®
concentration   10,000 mg/L Cd in nitric acid
application(s)   AAS: suitable
  ICP: suitable
format   single component solution
storage temp.   room temp

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the label. Information about trace impurities are stated on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Analysis Note

10′000 mg/L Cd in 5% nitric acid, prepared with high purity Cd, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Supplemental Hazard Statements 
EUH071
RIDADR 
UN 2031B 8 / PGII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?