EMAIL THIS PAGE TO A FRIEND
90244 Supelco

trans-2-Heptenal

analytical standard

Purchase

Properties

Related Categories 2009/48/EC (Directive on the Safety of Toys), A-L, Additional Standards, Aldehyde, All Alphabetically Sorted,
grade   analytical standard
vapor density   >1 (vs air)
InChI Key   NDFKTBCGKNOHPJ-AATRIKPKSA-N
assay   ≥95.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤8.0% water
refractive index   n20/D 1.450(lit.)
bp   90-91 °C/50 mmHg(lit.)
density   0.857 g/mL at 25 °C(lit.)
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Environmental
Food and Beverages
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1988 6.1(3) / PGIII
WGK Germany 
3
RTECS 
MJ8795000
Flash Point(F) 
123.8 °F
Flash Point(C) 
51 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?