EMAIL THIS PAGE TO A FRIEND
90728 Supelco

Ethylpyrazine

analytical standard

  • CAS Number 13925-00-3

  • Empirical Formula (Hill Notation) C6H8N2

  • Molecular Weight 108.14

  •  Beilstein/REAXYS Number 108200

  •  EC Number 237-691-5

  •  MDL number MFCD00006149

  •  PubChem Substance ID 329769945

  •  NACRES NA.24

Purchase

Properties

Related Categories A-F, A-L, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Analytical Standards,
Quality Level   100
grade   analytical standard
assay   ≥99.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.496-1.500
  n20/D 1.498 (lit.)
bp   152-153 °C (lit.)
density   0.984 g/mL at 25 °C (lit.)
suitability   complies for Proton NMR spectrum
Featured Industry   Cleaning Products
Cosmetics
Food and Beverages
Personal Care
format   neat
SMILES string   CCc1cnccn1
InChI   1S/C6H8N2/c1-2-6-5-7-3-4-8-6/h3-5H,2H2,1H3
InChI key   KVFIJIWMDBAGDP-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
RTECS 
UQ3330000
Flash Point(F) 
73.4 °F
Flash Point(C) 
23 °C
Protocols & Articles

Articles

GC Analysis of Pyrazines in Peanut Butter on SUPELCOWAX® 10 after SPME using 50/30 μm DVB/Carboxen/PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Pyrazines in Peanut Butter on SUPELCOWAX® 10 after SPME using 50/30 μm DVB/Carboxen/PDMS Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Separation, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Volatiles in Roasted Coffee Beans on Omegawax® after SPME using a 50/30 μm DVB/Carboxen/PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles in Roasted Coffee Beans on Omegawax® after SPME using a 50/30 μm DVB/Carboxen/PDMS Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?