• USA Home
  • 910235 - 2,3:4,5-Bis-O-(1-methylethylidene)-D-arabinose

EMAIL THIS PAGE TO A FRIEND
910235 Sigma-Aldrich

2,3:4,5-Bis-O-(1-methylethylidene)-D-arabinose

95%

Synonym: 2,3:4,5-Diisopropylidene-D-arabinose

  • CAS Number 13039-93-5

  • Empirical Formula (Hill Notation) C11H18O5

  • Molecular Weight 230.26

  •  MDL number MFCD15144988

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
assay   95%
form   liquid
refractive index   n/D 1.4477
density   1.1168 g/mL
storage temp.   2-8°C
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?