• USA Home
  • 91333 - FluoroSELECT Gram Negative Assay Kit

EMAIL THIS PAGE TO A FRIEND
91333 Millipore

FluoroSELECT Gram Negative Assay Kit

  •  NACRES NA.85

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Fluorescence/Luminescence Spectroscopy, FluoroSELECT Swab Tests, Handheld Fluorometers and Kits (FluoroSELECT),
Featured Industry   Food and Beverages
Quality Level   100
storage temp.   2-8°C
suitability   Escherichia coli
  algae
  coliforms

Description

General description

The FluoroSELECT product line combines portable, low cost, single-channel fluorometers with suitable kits for a wide range of applications.


Learn more about our FluoroSELECT

Components

Composition (for 50 assays):
Reagent A (Substrate): min. 4 mL
Reagent B (Enzyme Inducer): min. 2 mL
Reagent C (Lysing Agent): min. 6 mL
Incubation Media Powder: 0.4 g
Large Plastic Vials (Sample): 50 pcs
Small Plastic Vials (Mixing): 50 pcs
Mini Glass Tubes (Testing): 50 pcs.

Legal Information

FluoroSELECT is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
Flash Point(F) 
104 - 122 °F
Flash Point(C) 
40 - 50 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

FluoroSELECT™ – Handheld Fluorometers and Kits for Field and Lab Application

The FluoroSELECT™ product line combines portable, low cost, single-channel fluorometers with suitable kits for a wide range of applications. FluoroSELECT stands for highly sensitive and easy-to-use w...
Jens Boertz, Bernhard Schönenberger, Roland Meier
Analytix 2014, Issue 3
Keywords: Food & Beverage

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Related Products

related product

Product #

Description

Add to Cart

Z805602 FluoroSELECT single channel fluorometer, fluorescence λex 480 nm; λem 530 nm
Z805726 FluoroSELECT single channel fluorometer, fluorescence λex 360 nm; λem 460 nm
08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?