EMAIL THIS PAGE TO A FRIEND
91635 Supelco

Idaein chloride

analytical standard

Synonym: 3-(Galactosyloxy)-3′,4′,5,7-tetrahydroxyflavylium chloride, Cyanidin 3-O-β-D-galactopyranoside, Cyanidin 3-O-galactoside chloride, Idein chloride

  • CAS Number 27661-36-5

  • Empirical Formula (Hill Notation) C21H21ClO11

  • Molecular Weight 484.84

  •  MDL number MFCD00270387

  •  PubChem Substance ID 329770062

  •  NACRES NA.24

Purchase

Properties

Related Categories A - D, Alphabetical Index of Phytochemical Standards, Analytical Standards, Analytical/Chromatography, Flavonoids,
Quality Level   100
grade   analytical standard
assay   ≥95.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤10.0% Water (Karl Fischer)
Featured Industry   Food and Beverages
format   neat
shipped in   wet ice
storage temp.   −20°C
SMILES string   [Cl-].OC[C@H]1O[C@@H](Oc2cc3c(O)cc(O)cc3[o+]c2-c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@H]1O
InChI   1S/C21H20O11.ClH/c22-7-16-17(27)18(28)19(29)21(32-16)31-15-6-10-12(25)4-9(23)5-14(10)30-20(15)8-1-2-11(24)13(26)3-8;/h1-6,16-19,21-22,27-29H,7H2,(H3-,23,24,25,26);1H/t16-,17+,18+,19-,21-;/m1./s1
InChI key   YTMNONATNXDQJF-QSLGVYCOSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Anthocyanin.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?