EMAIL THIS PAGE TO A FRIEND
91975 Supelco

Mineral oil type A

for the determination of hydrocarbons (acc. to EN ISO 9377-2)

Synonym: Diesel oil without additives, Diesel oil

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, DIN EN ISO 9377-2,
form   neat
quality   for the determination of hydrocarbons (acc. to EN ISO 9377-2)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
Petroleum
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
NA 1993 / PGIII
WGK Germany 
2
Flash Point(F) 
176 °F
Flash Point(C) 
80 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?