• USA Home
  • 92473 - (±)-Naproxen-(methoxy-13C,d3)

EMAIL THIS PAGE TO A FRIEND
92473 Supelco

(±)-Naproxen-(methoxy-13C,d3)

analytical standard

Synonym: (RS)-2-[6-(Methoxy-13C,d3)-2-naphthyl]propionic acid, 6-(Methoxy-13C,d3)-α-methyl-2-naphthaleneacetic acid, [13C,2H3]-(±)-Naproxen

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography, Drugs & Metabolites,
grade   analytical standard
InChI Key   CMWTZPSULFXXJA-JVXUGDAPSA-N
assay   ≥98.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

LC-MS identity conforms

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?