EMAIL THIS PAGE TO A FRIEND
93508 Sigma-Aldrich

Dry ice

plates

Synonym: Carbon dioxide solid

  • CAS Number 124-38-9

  • Empirical Formula (Hill Notation) CO2

  • Molecular Weight 44.01

  •  Beilstein Registry Number 1900390

  •  MDL number MFCD00011491

Purchase

Properties

form   plates
shipped in   dry ice
storage temp.   −70°C

Description

Other Notes

Sales restrictions may apply

Safety & Documentation

Safety Information

RIDADR 
UN 1845PSN2 / PGIII
WGK Germany 
nwg
RTECS 
FF6400000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?