• USA Home
  • 94204 - tert-Butyl 3-oxopropionate

EMAIL THIS PAGE TO A FRIEND
94204 Sigma-Aldrich

tert-Butyl 3-oxopropionate

Synonym: tert-Butyl malonaldehydate

  • CAS Number 108350-21-6

  • Linear Formula (CH3)3COCOCH2CHO

  • Molecular Weight 144.17

Purchase

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?