EMAIL THIS PAGE TO A FRIEND
94575 Supelco

trans-2-Pentenal

analytical standard

Synonym: 2-Ethylacrylic aldehyde

Purchase

Properties

Related Categories Alcoholic & Nonalcoholic Beverages, Aldehyde, All Alphabetically Sorted, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   DTCCTIQRPGSLPT-ONEGZZNKSA-N
assay   ≥95.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤1.0% water
refractive index   n20/D 1.439-1.445
  n20/D 1.440-1.446(lit.)
bp   80-81 °C/160 mmHg(lit.)
density   0.86 g/mL at 25 °C(lit.)
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1989BE 3 / PGII
WGK Germany 
3
RTECS 
SB1560000
Flash Point(F) 
71.6 °F
Flash Point(C) 
22 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?