EMAIL THIS PAGE TO A FRIEND
94792 Millipore

Lactose Broth

for microbiology, ready to use media in the bottle

Purchase

Properties

Related Categories A - M, All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Liquid Media/Broths, Escherichia coli & Coliforms,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   lactose, 5 g/L
  meat extract, 3 g/L
  peptone, 5 g/L
final pH   6.9±0.2 (25 °C)
Featured Industry   Environmental
Food and Beverages
suitability   nonselective for Escherichia coli and Coliforms

Description

Application

Lactose Broth is a classical medium for use in the presumptive testing for coliforms and for the enrichment of Salmonella. This formulation is as per the standards recommended by APHA, AWWA, USP-NF and European Pharmacopoeia.

Safety & Documentation

Safety Information

WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?