• USA Home
  • 94898 - Vincristine sulfate salt

EMAIL THIS PAGE TO A FRIEND
94898 Sigma-Aldrich

Vincristine sulfate salt

≥95.0% (HPLC)

Synonym: 22-Oxovincaleukoblastine sulfate salt, Leurocristine sulfate salt, VCR

  • CAS Number 2068-78-2

  • Empirical Formula (Hill Notation) C46H56N4O10 · H2SO4

  • Molecular Weight 923.04

  •  Beilstein Registry Number 3924631

  •  EC Number 218-190-0

  •  MDL number MFCD00084729

Purchase

Properties

InChI Key   AQTQHPDCURKLKT-PNYVAJAMSA-N
assay   ≥95.0% (HPLC)
impurities   ≤10% water
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1/PG 2
WGK Germany 
3
RTECS 
OH6340000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

V8879 Vincristine sulfate salt, 95.0-105.0% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?