• USA Home
  • 96694 - Methotrexate-(methyl-13C,d3)

EMAIL THIS PAGE TO A FRIEND
96694 Supelco

Methotrexate-(methyl-13C,d3)

analytical standard

Synonym: [13C,2H3]-Methotrexate

  • Empirical Formula (Hill Notation) 13CC19D3H19N8O5

  • Molecular Weight 458.45

  •  MDL number MFCD27665512

  •  PubChem Substance ID 329770526

Purchase

Properties

Related Categories Additional Drugs, Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
InChI Key   FBOZXECLQNJBKD-AIFPBEHOSA-N
assay   ≥95.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat
storage temp.   2-8°C

Description

Analysis Note

LS-MS identity conforms

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?