EMAIL THIS PAGE TO A FRIEND
96709 Supelco

Oleamide

reference material

Synonym: (Ole), cis-9,10-Octadecenoamide, cis-9-Octadecenamide, Oleic acid amide

  • CAS Number 301-02-0

  • Empirical Formula (Hill Notation) C18H35NO

  • Molecular Weight 281.48

  •  MDL number MFCD00053638

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Chinese Food Contact Materials Regulation GB 9685 2016, Extractables and Leachables, Food & Beverage Standards,
grade   reference material
assay   ≥96.0% (GC)
shelf life   limited shelf life, expiry date on the label
storage temp.   −20°C
SMILES string   CCCCCCCC\C=C/CCCCCCCC(N)=O
InChI   1S/C18H35NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H2,19,20)/b10-9-
InChI key   FATBGEAMYMYZAF-KTKRTIGZSA-N

Description

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Biochem/physiol Actions

Sleep-inducing brain lipid, which allosterically modulates GABAA receptors and potentiates 5-HT7 serotonin receptor responses. Selective endogenous agonist of rat and human CB1 cannabinoid receptor.

Safety & Documentation

Safety Information

WGK Germany 
WGK 1
Flash Point(F) 
404.6 °F
Flash Point(C) 
207 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Reference Materials for Extractables and Leachables Testing

Extractables and Leachables (E&L) are chemical compounds with the potential to migrate into pharmaceutical or clinical products from packaging materials, tubing, or medical devices, resulting in pati...
Keywords: Addition reactions, Clinical, Gas chromatography, High performance liquid chromatography, Infrared spectroscopy, Ion-exchange chromatography, Nuclear magnetic resonance spectroscopy, Pharmaceutical

Related Products

replaced by

Product #

Description

Add to Cart

CRM96709 Oleamide, certified reference material, TraceCERT®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?