• USA Home
  • D0679 - 3,6-Diacetoxyphthalonitrile

EMAIL THIS PAGE TO A FRIEND
D0679 Sigma-Aldrich

3,6-Diacetoxyphthalonitrile

Synonym: 1,4-Diacetoxy-2,3-dicyanobenzene, 2,3-Dicyano-1,4-hydroquinone diacetate, ADB

  • CAS Number 83619-73-2

  • Empirical Formula (Hill Notation) C12H8N2O4

  • Molecular Weight 244.20

  •  Beilstein/REAXYS Number 2874970

  •  MDL number MFCD00036857

  •  PubChem Substance ID 24893285

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Cell Biology, D, Fluorescent Indicators and Probes,
mp   166-167 °C (lit.)
storage temp.   −20°C
SMILES string   CC(=O)Oc1ccc(OC(C)=O)c(C#N)c1C#N
InChI   1S/C12H8N2O4/c1-7(15)17-11-3-4-12(18-8(2)16)10(6-14)9(11)5-13/h3-4H,1-2H3
InChI key   VXDCGBVAZDYTFK-UHFFFAOYSA-N

Description

Application

for intracellular pH measurements using FCM

Substrates

Esterase substrate

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Sensor Dyes

In 1998 R.F. Furchgott, L.J. Ignarro, and F. Murad received the Nobel Prize in Physiology and Medicine for their discoveries concerning nitric oxide as a signaling molecule in the cardiovascular syst...
Monika Baeumle
BioFiles 2011, 6.3, 23.
Keywords: Applications, Biochemistry, Biological processes, Cancer, Cardiovascular, Central Nervous System, Diseases, Extinction coefficient, Indicators, Inflammation, Methods, Microscopy, Neurotransmission, Sol-gel, Type

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?