• USA Home
  • D1053 - Dextromethorphan hydrobromide monohydrate

EMAIL THIS PAGE TO A FRIEND
D1053 Sigma-Aldrich

Dextromethorphan hydrobromide monohydrate

analytical standard

Synonym: (9S,13S,14S)-3-Methoxy-17-methylmorphinan hydrobromide

  • CAS Number 6700-34-1

  • Empirical Formula (Hill Notation) C18H25NO · HBr · H2O

  • Molecular Weight 370.32

  •  Beilstein/REAXYS Number 6453793

  •  MDL number MFCD02173901

  •  PubChem Substance ID 24893336

Purchase

Properties

grade   analytical standard
packaging   vial of 2 g
SMILES string   Br[H].[H]O[H].[H][C@]12CCCC[C@]13CCN(C)[C@H]2Cc4ccc(OC)cc34
InChI   1S/C18H25NO.BrH.H2O/c1-19-10-9-18-8-4-3-5-15(18)17(19)11-13-6-7-14(20-2)12-16(13)18;;/h6-7,12,15,17H,3-5,8-11H2,1-2H3;1H;1H2/t15-,17+,18+;;/m1../s1
InChI key   STTADZBLEUMJRG-IKNOHUQMSA-N

Description

Packaging

Supplied in amber screw-cap vials

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Methorphan Enantiomers on Astec® CHIROBIOTIC® V2

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Methorphan Enantiomers on Astec® CHIROBIOTIC® V2
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?