EMAIL THIS PAGE TO A FRIEND
D9689 Sigma-Aldrich

DABCYL SE

solid

Synonym: 4-([4-(Dimethylamino)phenyl]­azo)­benzoic acid succinimidyl ester

  • Empirical Formula (Hill Notation) C19H18N4O4

  • Molecular Weight 366.37

  •  MDL number MFCD00467415

  •  PubChem Substance ID 24894321

Purchase

Properties

Related Categories Cell Biology, D, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
form   solid
mol wt   apparent mol wt 366.38
shipped in   dry ice
storage temp.   −20°C
SMILES string   CN(C)c1ccc(cc1)\N=N\c2ccc(cc2)C(=O)ON3C(=O)CCC3=O
InChI   1S/C19H18N4O4/c1-22(2)16-9-7-15(8-10-16)21-20-14-5-3-13(4-6-14)19(26)27-23-17(24)11-12-18(23)25/h3-10H,11-12H2,1-2H3/b21-20+
InChI key   IBOVDNBDQHYNJI-QZQOTICOSA-N

Description

Application

Non-fluorescent acceptor in fluorescent resonance energy transfer applications.

This succinimidyl ester is useful for preparation of quenched fluorogenic substrates for proteases.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?