EMAIL THIS PAGE TO A FRIEND
F9676 Sigma-Aldrich

Fmoc Biocytin

Synonym: ε-Biotinoyl-α-(9-fluorenylmethoxycarbonyl)-L-lysine

  • Empirical Formula (Hill Notation) C31H38N4O6S

  • Molecular Weight 594.72

  •  MDL number MFCD00270741

  •  PubChem Substance ID 24895000

Purchase

Properties

mol wt   594.72
solubility   DMF: soluble
  acetonitrile: soluble
shipped in   wet ice
storage temp.   −20°C
SMILES string   OC(=O)[C@H](CCCCNC(=O)CCCC[C@@H]1SCC2NC(=O)NC12)NC(=O)OCC3c4ccccc4-c5ccccc35

Description

Application

Useful in automated synthesis of biotinylated peptides.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?