EMAIL THIS PAGE TO A FRIEND
G0289 Millipore

Gelatin iron medium

for microbiology

Purchase

Description

Components

Ingredients (g/L)
Peptic digest of animal tissue, 25.00
Meat extract, 7.50
Sodium chloride, 5.00
Gelatin, 120.00
Ferrous chloride, 0.50
Agar, 1.00

Application

Used for detecting gelatin liquification and hydrogen sulfide production.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Clostridium perfringens, their Properties and their Detection

Clostridia are relatively large, Gram-positive, rod-shaped bacteria that can undergo only anaerobic metabolism. Most Clostridia cannot grow under aerobic conditions and can even be killed by exposure...
ivo.siegrist@sial.com
AnalytiX Volume 10 Article 4
Keywords: Aerobic, Anaerobic, Chlorinations, Degradations, Environmental, Fermentation, Germination, Gram stain, Indicators, Metabolism, Microbiology

EZ-Product Family for Bioburden Testing

The EZ family has been designed to optimize and streamline your bioburden workflow. Ideal for microbiological water, beverage, and medical device testing using the membrane filtration method. The fam...
Keywords: Filtration

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?