• USA Home
  • H6404 - Hexamethylenetetramine

EMAIL THIS PAGE TO A FRIEND
H6404 Sigma-Aldrich

Hexamethylenetetramine

BioXtra

Synonym: 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, Hexamine, Methenamine, Urotropine

  • CAS Number 100-97-0

  • Empirical Formula (Hill Notation) C6H12N4

  • Molecular Weight 140.19

  •  Beilstein/REAXYS Number 2018

  •  EC Number 202-905-8

  •  MDL number MFCD00006895

  •  PubChem Substance ID 24895743

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, N-Containing, Other Heterocyclic Building Blocks More...
vapor pressure   <0.01 mmHg ( 20 °C)
product line   BioXtra
autoignition temp.   770 °F
impurities   <0.0005% Phosphorus (P)
  <0.1% Insoluble matter
ign. residue   <0.1%
mp   280 °C (subl.) (lit.)
solubility   1 M acetic acid: 0.5 M, clear, colorless
anion traces   chloride (Cl-): <0.05%
  sulfate (SO42-): 0.05%
cation traces   Al: <0.0005%
  Ca: <0.001%
  Cu: <0.0005%
  Fe: <0.0005%
  K: <0.005%
  Mg: <0.0005%
  NH4+: <0.05%
  Na: <0.01%
  Pb: <0.001%
  Zn: <0.0005%
SMILES string   C1N2CN3CN1CN(C2)C3
InChI   1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2
InChI key   VKYKSIONXSXAKP-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1328 - class 4.1 - PG 3 - Hexamethylenetetramine
WGK Germany 
1
RTECS 
MN4725000
Flash Point(F) 
482 °F
Flash Point(C) 
250 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?