EMAIL THIS PAGE TO A FRIEND
I8136 Sigma-Aldrich

Iodoacetic acid

~99% (titration), powder

  • CAS Number 64-69-7

  • Linear Formula ICH2CO2H

  • Molecular Weight 185.95

  •  Beilstein Registry Number 1739079

  •  EC Number 200-590-1

  •  MDL number MFCD00002685

Purchase

Properties

InChI Key   JDNTWHVOXJZDSN-UHFFFAOYSA-N
assay   ~99% (titration)
form   powder
packaging   ampule of 500 mg
color   white to off-white
mp   77-79 °C (lit.)
storage temp.   −20°C

Description

Application

Reagent for the modification of cysteine residues in proteins.

Packaging

Packaged under argon in sealed ampules

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2923 6.1(8) / PGI
WGK Germany 
1
RTECS 
AI3500000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I4386 Iodoacetic acid, ≥98.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?