• USA Home
  • P1528 - Pararosaniline hydrochloride

EMAIL THIS PAGE TO A FRIEND
P1528 Sigma-Aldrich

Pararosaniline hydrochloride

certified by the Biological Stain Commission

Synonym: Basic Fuchsin, Basic Parafuchsin, Basic Red 9, Magenta O, Parafuchsin hydrochloride, Paramagenta hydrochloride, Pararosaniline chloride

  • CAS Number 569-61-9

  • Linear Formula C19H18N3Cl

  • Molecular Weight 323.82

  •  Colour Index Number 42500

  •  Beilstein/REAXYS Number 4164603

  •  EC Number 209-321-2

  •  MDL number MFCD00001657

  •  PubChem Substance ID 24898215

Purchase

Properties

Related Categories Cell Biology, Certified Stains & Dyes, Hematology and Histology, P, Stains & Dyes, A to Z,
grade   certified by the Biological Stain Commission
composition   Dye content, ≥88%
SMILES string   Cl.Nc1ccc(cc1)C(\c2ccc(N)cc2)=C3/C=CC(=N)C=C3
InChI   1S/C19H17N3.ClH/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15;/h1-12,20H,21-22H2;1H
InChI key   JUQPZRLQQYSMEQ-UHFFFAOYSA-N

Description

Other Notes

This is the pure, principal component of mixtures commonly referred to as Basic Fuchsin.

Suitability

Certified for use in acid-fast staining procedures utilizing carbol-fuchsin; Endo medium; as Schiff′s reagent in the Periodic Acid Schiff (PAS) and Feulgen techniques; Gomori′s aldehyde fuchsin method for pancreatic B cells and hepatitis B surface antigen.

Legal Information

Magenta is a trademark of Magenta Corp.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
CX9850100
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?