• USA Home
  • T0199 - Trichloroacetic acid solution

EMAIL THIS PAGE TO A FRIEND
T0199 Sigma-Aldrich

Trichloroacetic acid solution

0.6 N

Synonym: TCA

  • CAS Number 76-03-9

  • Linear Formula Cl3CCOOH

  • Molecular Weight 163.39

  •  Beilstein/REAXYS Number 970119

  •  MDL number MFCD00004177

  •  PubChem Substance ID 329826591

Purchase

Properties

concentration   ~10 % (w/v)
SMILES string   OC(=O)C(Cl)(Cl)Cl
InChI   1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)
InChI key   YNJBWRMUSHSURL-UHFFFAOYSA-N

Description

Application

Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation.
Used as a decalcifier and fixative in microscopy.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Respiratory system
RIDADR 
UN 2564 8 / PGII
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?