• USA Home
  • T5883 - Sodium triphosphate pentabasic

EMAIL THIS PAGE TO A FRIEND
T5883 Sigma-Aldrich

Sodium triphosphate pentabasic

practical grade, 90-95%

Synonym: Pentasodium tripolyphosphate, Sodium tripolyphosphate pentabasic

Purchase

Properties

grade   practical grade
assay   90-95%
SMILES string   [Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O
InChI   1S/5Na.H5O10P3/c;;;;;1-11(2,3)9-13(7,8)10-12(4,5)6/h;;;;;(H,7,8)(H2,1,2,3)(H2,4,5,6)/q5*+1;/p-5
InChI key   HWGNBUXHKFFFIH-UHFFFAOYSA-I

Description

Other Notes

Straight chain phosphates

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
1
RTECS 
YK4570000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?