EMAIL THIS PAGE TO A FRIEND
A178 Supelco

Acetohexamide

analytical standard

Synonym: 4-Acetyl-N-[(cyclohexylamino)carbonyl]benzenesulfonamide

Purchase

Properties

Related Categories Additional Drugs, Analytical Standards, Analytical/Chromatography, Pharmaceuticals, Illicit Drugs & Alcohol
Quality Level   100
grade   analytical standard
form   neat
  solid
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
color   white
solubility   DMSO: ~45 mg/mL
  H2O: insoluble
Featured Industry   Forensics and Toxicology
Pharmaceutical (small molecule)
format   neat
SMILES string   CC(=O)c1ccc(cc1)S(=O)(=O)NC(=O)NC2CCCCC2
InChI   1S/C15H20N2O4S/c1-11(18)12-7-9-14(10-8-12)22(20,21)17-15(19)16-13-5-3-2-4-6-13/h7-10,13H,2-6H2,1H3,(H2,16,17,19)
InChI key   VGZSUPCWNCWDAN-UHFFFAOYSA-N
Gene Information   human ... ABCC8(6833), KCNJ1(3758), KCNJ11(3767)

Description

Application

Acetohexamide may be used as a reference standard for the determination of acetohexamide in presence of 1-methylnicotinamide reagent in plasma sample and tablet formulations by spectrofluorimetry method.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

1 g in poly bottle

Biochem/physiol Actions

Oral hypoglycemic agent

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. A178.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
RTECS 
YR7350000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?