EMAIL THIS PAGE TO A FRIEND
A2039 Sigma-Aldrich

Aluminum oxide

Type A-5

Synonym: Alumina

Purchase

Properties

Related Categories Aluminum, Aluminum Catalysts, Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science,
type   Type A-5
mp   2040 °C (lit.)
SMILES string   O=[Al]O[Al]=O
InChI   1S/2Al.3O
InChI key   TWNQGVIAIRXVLR-UHFFFAOYSA-N

Description

Application

For disintegrating yeast cells and microorganisms by grinding or blending.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
BD1200000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?