EMAIL THIS PAGE TO A FRIEND
ARK2163 SAFC

Benzyl alcohol

PharmaGrade, USP/NF, Ph. Eur., Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.

Synonym: Benzenemethanol

  • CAS Number 100-51-6

  • Linear Formula C6H5CH2OH

  • Molecular Weight 108.14

  •  Beilstein/REAXYS Number 878307

  •  EC Number 202-859-9

  •  MDL number MFCD00004599

  •  PubChem Substance ID 329773052

  •  NACRES NA.21

Purchase

Properties

Quality Level   500
biological source   synthetic
Agency/Method   Ph. Eur.
  USP/NF
vapor density   3.7 (vs air)
vapor pressure   13.3 mmHg ( 100 °C)
  3.75 mmHg ( 77 °C)
product line   PharmaGrade
form   liquid
autoignition temp.   817 °F
shelf life   2 yr
refractive index   n20/D 1.539 (lit.)
bp   203-205 °C (lit.)
mp   −16-−13 °C (lit.)
density   1.045 g/mL at 25 °C (lit.)
suitability   suitable for manufacturing use
shipped in   wet ice
storage temp.   2-8°C
SMILES string   OCc1ccccc1
InChI   1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2
InChI key   WVDDGKGOMKODPV-UHFFFAOYSA-N

Description

General description

PharmaGrade products are manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production. For more information on the PharmaGrade Program, visit sigma-aldrich.com/safc-pharmagrade.

Linkage

Enhanced Quality Product Documentation on PharmaGrade Benzyl Alcohol

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3334
WGK Germany 
WGK 1
RTECS 
DN3150000
Flash Point(F) 
213.8 °F - DIN 51758
Flash Point(C) 
101 °C - DIN 51758
Protocols & Articles

Related Content

Enhanced Quality Program [VIDEO]

The Enhanced Quality Program is a quality management system that is used to differentiate products based on the required level of transparency, control and documentation support determined by the nee...

One Quality Policy [VIDEO]

Sigma-Aldrich maintains one quality policy on a global level to provide consistency in vision, maintained across all manufacturing and distribution sites so that the company on the whole is adhering ...

Quality Management Systems [VIDEO]

See how we combine the use of various Quality Management Systems to manage manufacturing and supply chain for our products, as well as ensure data integrity for information provided to customers via ...

Risk Management [VIDEO]

Our Quality Policy means that we are able to provide certain information to end customers based on a product’s Enhanced Quality level, and that information is limited to what we are able to guarantee...

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

1.37120 Benzyl alcohol, EMPROVE® EXPERT, Ph. Eur., ChP, JP, NF

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?