• USA Home
  • B4386 - Benoxinate hydrochloride

EMAIL THIS PAGE TO A FRIEND
B4386 Sigma-Aldrich

Benoxinate hydrochloride

≥98%

Synonym: 2-(Diethylamino)ethyl 4-amino-3-butoxybenzoate hydrochloride, 4-Amino-3-butoxybenzoic acid diethylaminoethyl ester, Oxybuprocaine hydrochloride

Purchase

Properties

assay   ≥98%
SMILES string   Cl[H].CCCCOc1cc(ccc1N)C(=O)OCCN(CC)CC
InChI   1S/C17H28N2O3.ClH/c1-4-7-11-21-16-13-14(8-9-15(16)18)17(20)22-12-10-19(5-2)6-3;/h8-9,13H,4-7,10-12,18H2,1-3H3;1H
InChI key   PRGUDWLMFLCODA-UHFFFAOYSA-N
Gene Information   human ... SCN10A(6336), SCN11A(11280), SCN1A(6323), SCN2A(6326), SCN3A(6328), SCN4A(6329), SCN5A(6331), SCN7A(6332), SCN8A(6334), SCN9A(6335)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
DG1502900

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B9050 Benoxinate hydrochloride, meets USP testing specifications

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?