EMAIL THIS PAGE TO A FRIEND
B6638 Supelco

Butalbital

Synonym: 5-Allyl-5-isobutylbarbituric acid

  • CAS Number 77-26-9

  • Empirical Formula (Hill Notation) C11H16N2O3

  • Molecular Weight 224.26

  •  Beilstein/REAXYS Number 202119

  •  EC Number 201-017-8

  •  MDL number MFCD00056089

  •  PubChem Substance ID 329773281

Purchase

Properties

form   neat
drug control   USDEA Schedule III; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
SMILES string   CC(C)CC1(CC=C)C(=O)NC(=O)NC1=O
InChI   1S/C11H16N2O3/c1-4-5-11(6-7(2)3)8(14)12-10(16)13-9(11)15/h4,7H,1,5-6H2,2-3H3,(H2,12,13,14,15,16)
InChI key   UZVHFVZFNXBMQJ-UHFFFAOYSA-N
Gene Information   human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Sedative/hypnotic

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
CP8750000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Barbiturates on Discovery® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Discovery® C18
Keywords: Chromatography, High performance liquid chromatography

UHPLC Analysis of Barbiturates on Titan™ C18

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Barbiturates on Titan™ C18
Keywords: Anti-depressants, Anticonvulsants, Chromatography, Clinical, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pharmaceutical, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?