• USA Home
  • BCR321 - Unalloyed zinc (trace elements)

EMAIL THIS PAGE TO A FRIEND
BCR321 Sigma-Aldrich

Unalloyed zinc (trace elements)

BCR® certified Reference Material

Synonym: Zinc, unalloyed

Purchase

Description

Analysis Note

For more information please see:
BCR321

Legal Information

BCR is a registered trademark of European Commission

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?