EMAIL THIS PAGE TO A FRIEND
C1653 Supelco

Codeine solution

analytical standard, for drug analysis, 1.0 mg/mL in methanol

Purchase

Properties

Related Categories β-Glucuronidase Drug Analysis Bundle, 1694 Pharmaceuticals & Personal Care Products, Additional Standards, Analytical Standards, Analytical and Industrial Enzymes,
grade   analytical standard, for drug analysis
Agency/Method   EPA 1694
drug control   Home Office Schedule 2; stupéfiant (France)
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
Pharmaceutical (small molecule)
format   single component solution
storage temp.   2-8°C
SMILES string   [H][C@@]12Oc3c(OC)ccc4C[C@H]5N(C)CC[C@@]1(c34)[C@@]5([H])C=C[C@@H]2O
InChI   1S/C18H21NO3/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19/h3-6,11-13,17,20H,7-9H2,1-2H3/t11-,12+,13-,17-,18-/m0/s1
InChI key   OROGSEYTTFOCAN-DNJOTXNNSA-N
Gene Information   human ... CYP2D6(1565), OPRD1(4985), OPRK1(4986), OPRM1(4988)
rat ... Cyp2d1(266684), Cyp2d2(25053), Cyp2d3(24303), Cyp2d4v1(171522)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

1 mL in ampule

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. C1653.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles

Articles

GC Analysis of Basic Drugs on Equity®-5

From our library of Articles, Sigma-Aldrich presents GC Analysis of Basic Drugs on Equity®-5
Keywords: Chromatography, Gas chromatography, Purification

GC Analysis of Basic Drugs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after SPME using 65 μm PDMS/DVB Fiber, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Basic Drugs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after SPME using 65 μm PDMS/DVB Fiber, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification, Solid phase microextractions

GC Analysis of Basic Drugs on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Basic Drugs on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

HPLC Analysis of Alkaloids on Discovery® RP-AmideC16

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Alkaloids on Discovery® RP-AmideC16
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Alkaloids on Discovery® Zr-PBD

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Alkaloids on Discovery® Zr-PBD
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical, Vitamins

HPLC Analysis of Amides on Discovery® RP-Amide C16

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Amides on Discovery® RP-Amide C16
Keywords: Analgesics, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Analgesics on Discovery® RP-Amide C16

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Analgesics on Discovery® RP-Amide C16
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Analgesics, Narcotics on Ascentis® Phenyl

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Analgesics, Narcotics on Ascentis® Phenyl
Keywords: Analgesics, Chromatography, Clinical, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Antitussive/Antihistamine/Antipyretic Mix on Discovery® RP-Amide C16

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Antitussive/Antihistamine/Antipyretic Mix on Discovery® RP-Amide C16
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Codeine and Metabolite on Ascentis® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Codeine and Metabolite on Ascentis® C18
Keywords: Analgesics, Chromatography, Clinical, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Codeine and Metabolite on Ascentis® RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Codeine and Metabolite on Ascentis® RP-Amide
Keywords: Analgesics, Chromatography, Clinical, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Cold Remedy Components on Ascentis® Express 2.0 μm RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Cold Remedy Components on Ascentis® Express 2.0 μm RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical, Separation

HPLC Analysis of Drugs of Abuse on Ascentis® Express F5

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Drugs of Abuse on Ascentis® Express F5
Keywords: Chromatography, High performance liquid chromatography

HPLC Analysis of Drugs of Abuse on Ascentis® Phenyl

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Drugs of Abuse on Ascentis® Phenyl
Keywords: Chromatography, High performance liquid chromatography

LC-MS Analysis of Pain Management Opiates using Ascentis® Express Biphenyl

From our library of Articles, Sigma-Aldrich presents LC-MS Analysis of Pain Management Opiates using Ascentis® Express Biphenyl
Keywords: Chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pharmaceutical, Separation

LC-MS Anlaysis of Pain Managment Opiates on Ascentis® Express Biphenyl, 30 x 2.1 mm

From our library of Articles, Sigma-Aldrich presents LC-MS Anlaysis of Pain Managment Opiates on Ascentis® Express Biphenyl, 30 x 2.1 mm
Keywords: Chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pharmaceutical, Separation

LC/MS (TOF) Analysis of Opioid Drugs and Glucuronide Metabolites in Urine on Ascentis® Express HILIC after SPE using Supel™-Select HLB, β-Glucuronidase Enzyme Digestion

From our library of Articles, Sigma-Aldrich presents LC/MS (TOF) Analysis of Opioid Drugs and Glucuronide Metabolites in Urine on Ascentis® Express HILIC after SPE using Supel™-Select HLB, β-Glucuronidase Enzyme Digestion
Keywords: Chromatography, High performance liquid chromatography, Mass spectrometry, Metabolites, Sample preparations, Solid phase extractions

LC/MS (TOF) Analysis of Opioids and Opiate-Dependence Management Drugs on Ascentis® Express HILIC

From our library of Articles, Sigma-Aldrich presents LC/MS (TOF) Analysis of Opioids and Opiate-Dependence Management Drugs on Ascentis® Express HILIC
Keywords: Chromatography, Clinical, High performance liquid chromatography, Mass spectrometry

LC/MS Analysis of Opioid Glucuronide Metabolites in Urine on Ascentis® Express F5 after Solid Phase Extraction (SPE) using Supel™-Select HLB

From our library of Articles, Sigma-Aldrich presents LC/MS Analysis of Opioid Glucuronide Metabolites in Urine on Ascentis® Express F5 after Solid Phase Extraction (SPE) using Supel™-Select HLB
Keywords: Chromatography, Clinical, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Metabolites, Sample preparations, Separation, Solid phase extractions, Solvents

LC/MS/MS Analysis of Drugs in Plasma on Ascentis® Express C18 after Extraction using SPME LC Tips

From our library of Articles, Sigma-Aldrich presents LC/MS/MS Analysis of Drugs in Plasma on Ascentis® Express C18 after Extraction using SPME LC Tips
Keywords: Chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Sample preparations, Separation, Solid phase microextractions

LC/MS/MS Analysis of Drugs of Abuse: Generic Screening on Ascentis® Express F5

From our library of Articles, Sigma-Aldrich presents LC/MS/MS Analysis of Drugs of Abuse: Generic Screening on Ascentis® Express F5
Keywords: Chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Solvents

TLC Analysis of Analgesics on HPTLC Pre-coated Plate RP-18 WF254s (analyte set 1)

From our library of Articles, Sigma-Aldrich presents TLC Analysis of Analgesics on HPTLC Pre-coated Plate RP-18 WF254s (analyte set 1)
Keywords: Pharmaceutical, Thin layer chromatography

TLC Analysis of Drugs on HPTLC Silica Gel 60 RP-18 F254s

From our library of Articles, Sigma-Aldrich presents TLC Analysis of Drugs on HPTLC Silica Gel 60 RP-18 F254s
Keywords: Chromatography, Pharmaceutical, Thin layer chromatography

UHPLC/MS Analysis of Heroin and Related Compounds on Titan™ C18

From our library of Articles, Sigma-Aldrich presents UHPLC/MS Analysis of Heroin and Related Compounds on Titan™ C18
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pharmaceutical, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?