• USA Home
  • C200 - Flow Cup Viscosity Standard

EMAIL THIS PAGE TO A FRIEND
C200

Flow Cup Viscosity Standard

UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 557.7 mPa.s (20 °C)

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Flow Cup Viscosity Standards, Paragon Scientific, Physical Properties Standards,
shelf life   limited shelf life, expiry date on the label
mfr. no.   Paragon Scientific Ltd
viscosity   557.7 mPa.s(20 °C)
Featured Industry   Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care
format   mixture

Description

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Flow Cup Viscosity Standards can be found on the Paragon Scientific webpage

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>199.4 °F
Flash Point(C) 
>93 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?