EMAIL THIS PAGE TO A FRIEND
C2155000

Cholesterol

European Pharmacopoeia (EP) Reference Standard

Synonym: 3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol

  • CAS Number 57-88-5

  • Empirical Formula (Hill Notation) C27H46O

  • Molecular Weight 386.65

  •  Beilstein/REAXYS Number 1915888

  •  MDL number MFCD00003646

  •  PubChem Substance ID 329774968

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, EP Standards, EP Standards C - D, Pharmacopeia & Metrological Institutes Standards More...
biological source   ovine
grade   pharmaceutical primary standard
Agency/Method   EP Reference Standard
form   neat
mfr. no.   EDQM
bp   360 °C (lit.)
mp   147-149 °C (lit.)
density   1.067 g/mL at 25 °C (lit.)
Featured Industry   Pharmaceutical (small molecule)
format   neat
SMILES string   CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
InChI   1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1
InChI key   HVYWMOMLDIMFJA-DPAQBDIFSA-N

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

Packaging

Unit quantity: 80 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference subtance catalogue.

Caution

Please find SDS provided by EDQM here..

Other Notes

Sales restrictions may apply.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of Sterols (Silylated Derivatives) in Olive Oil (Free Sterol Fraction) on SLB®-5ms after SPE using Discovery DSC-Si

From our library of Articles, Sigma-Aldrich presents GC Analysis of Sterols (Silylated Derivatives) in Olive Oil (Free Sterol Fraction) on SLB®-5ms after SPE using Discovery DSC-Si
Keywords: Agriculture, Calorimetry, Chromatography, Fractionation, Gas chromatography, Gas chromatography mass spectrometry, Mass selective detector, Mass spectrometry, Purification, Sample preparations, Saponifications, Silylations, Solid phase extractions

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C
PHR1533 Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?