EMAIL THIS PAGE TO A FRIEND
CRM113 Supelco

BNAs - Loamy Sand 1

certified reference material, pkg of 100 g

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Environmental Matrix CRMs (RTC),
grade   certified reference material
packaging   pkg of 100 g
mfr. no.   RTC, CRM113-100
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
pH   7.95
Featured Industry   Environmental
format   matrix material
storage temp.   2-8°C

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis. Download your certificate of analysis at: http://www.sigma-aldrich.com.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Specifications

View example COA for CRM113

Note - The link provided is an example certificate of analysis (COA) only and may not be the lot that you receive. All analytes may not be present or certified for each lot, and analyte concentrations will vary by lot.

Analyte

Description

Product #

Add to Cart

Benzo[b]fluoranthene    
Benzo[a]pyrene    
Bis(2-ethylhexyl) phthalate    
Chrysene    
2,4-Dinitrotoluene    
Fluoranthene    
Fluorene    
Hexachlorobenzene    
Hexachloroethane    
2-Nitroaniline    
3-Nitroaniline    
Nitrobenzene    
Pyrene    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?